siRNA Details
| Gene Symbol: LOC101928517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110732.1 | 5 | 1461 | |||
| GenBank mRNA | BC045766.1 | - | 5 | 1462 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928517| Gene Symbol: LOC101928517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110732.1 | 5 | 1461 | |||
| GenBank mRNA | BC045766.1 | - | 5 | 1462 | ||