siRNA Details
Gene Symbol: LOC101929698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110619.1 | 1, 2 | 704 | |||
GenBank mRNA | AK097865.1 | - | 1, 2 | 704 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110619.1 | 1, 2 | 704 | |||
GenBank mRNA | AK097865.1 | - | 1, 2 | 704 |