siRNA Details
| Gene Symbol: FGFR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK001052.1 | - | 1 | 226 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AU118968.1 | - | 1 | 226 | Chr. 8 - 38434840 - 38435664 [-] on Build GRCh38 | |

FGFR1
Genomic Map