siRNA Details
| Gene Symbol: MCM8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110101.1 | 5 | 603 | |||
| GenBank mRNA | AK058185.1 | - | 5 | 603 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MCM8-AS1| Gene Symbol: MCM8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110101.1 | 5 | 603 | |||
| GenBank mRNA | AK058185.1 | - | 5 | 603 | ||