siRNA Details
Gene Symbol: MCM8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110101.1 | 5 | 849 | |||
GenBank mRNA | AK058185.1 | - | 5 | 849 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MCM8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110101.1 | 5 | 849 | |||
GenBank mRNA | AK058185.1 | - | 5 | 849 |