siRNA Details
| Gene Symbol: C14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021390.1 | XP_016876879.1 | 1 | 230 | ||
| GenBank mRNA | AK024794.1 | - | 1 | 1139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14orf119| Gene Symbol: C14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021390.1 | XP_016876879.1 | 1 | 230 | ||
| GenBank mRNA | AK024794.1 | - | 1 | 1139 | ||