siRNA Details
| Gene Symbol: C14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017924.3 | NP_060394.1 | 1 | 91 | ||
| GenBank mRNA | AF061731.1 | - | 1 | 61 | ||
| AK024794.1 | - | 1 | 845 | |||
| BC056255.1 | - | 1 | 62 | 4694305 | ||
| BC061637.1 | - | 1 | 26 | 5532778 | ||
| DA605680.1 | - | 1 | 91 | |||

C14orf119
Genomic Map