siRNA Details
| Gene Symbol: C14orf119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017924.3 | NP_060394.1 | 1 | 60 | ||
| GenBank mRNA | AF061731.1 | - | 1 | 30 | ||
| AK024794.1 | - | 1 | 814 | |||
| BC056255.1 | - | 1 | 31 | 4694305 | ||
| DA605680.1 | - | 1 | 60 | |||

C14orf119
Genomic Map