siRNA Details
| Gene Symbol: LOC101929002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135253.1 | 2 | 1087 | |||
| GenBank mRNA | BC042084.1 | - | 2 | 1087 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555299.1 | - | 2 | 1087 | Chr. 14 - 92905697 - 92907482 [-] on Build GRCh38 | |

LOC101929002
Genomic Map