siRNA Details
| Gene Symbol: LOC101928583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135555.1 | 3 | 2100 | |||
| GenBank mRNA | BC042089.1 | - | 2 | 2261 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928583| Gene Symbol: LOC101928583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135555.1 | 3 | 2100 | |||
| GenBank mRNA | BC042089.1 | - | 2 | 2261 | ||