siRNA Details
| Gene Symbol: RASAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000981.1 | - | 1 | 613 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AU118091.1 | - | 1 | 613 | Chr. 1 - 178143886 - 178144723 [+] on Build GRCh38 | |

RASAL2
Genomic Map