siRNA Details
| Gene Symbol: LINC01128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047519.1 | 6 | 1687 | |||
| NR_047521.1 | 5 | 1534 | ||||
| NR_047523.1 | 4 | 1438 | ||||
| NR_047524.1 | 4 | 1350 | ||||
| NR_047525.1 | 5 | 1401 | ||||
| GenBank mRNA | AK026873.1 | - | 7 | 1285 | ||
| BC042880.1 | - | 5 | 1516 | |||

LINC01128
Genomic Map