siRNA Details
| Gene Symbol: ZBTB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 26847 | ||
| NM_001164343.2 | NP_001157815.1 | 12 | 27309 | |||
| NM_001164344.2 | NP_001157816.1 | 8 | 27273 | |||
| NM_001164345.2 | NP_001157817.1 | 7 | 27227 | |||
| NM_001164346.2 | NP_001157818.1 | 6 | 26866 | |||
| NM_001164347.2 | NP_001157819.1 | 6 | 26888 | |||
| NM_015642.5 | NP_056457.3 | 10 | 27071 | |||
| NR_121662.1 | 7 | 25329 | ||||
| GenBank mRNA | AL049423.1 | - | 1 | 1625 | ||

ZBTB20
Genomic Map