siRNA Details
| Gene Symbol: ZBTB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 25627 | ||
| NM_001164343.2 | NP_001157815.1 | 12 | 26089 | |||
| NM_001164344.2 | NP_001157816.1 | 8 | 26053 | |||
| NM_001164345.2 | NP_001157817.1 | 7 | 26007 | |||
| NM_001164346.2 | NP_001157818.1 | 6 | 25646 | |||
| NM_001164347.2 | NP_001157819.1 | 6 | 25668 | |||
| NM_015642.5 | NP_056457.3 | 10 | 25851 | |||
| NR_121662.1 | 7 | 24109 | ||||
| GenBank mRNA | AL049423.1 | - | 1 | 398 | ||
| BC035518.1 | - | 1 | 855 | |||

ZBTB20
Genomic Map