siRNA Details
| Gene Symbol: ZSCAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005249296.4 | XP_005249353.1 | 1 | 2431 | ||
| XM_011514807.2 | XP_011513109.1 | 1 | 2431 | |||
| XM_011514808.2 | XP_011513110.1 | 1 | 2431 | |||
| XM_017011196.1 | XP_016866685.1 | 1 | 2431 | |||
| GenBank mRNA | AK123280.1 | - | 1 | 500 | ||

ZSCAN31
Genomic Map