siRNA Details
| Gene Symbol: DAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521414.1 | XP_011519716.1 | 11 | 2417 | ||
| XM_017022044.1 | XP_016877533.1 | 11 | 2413 | |||
| GenBank mRNA | AK094290.1 | - | 1 | 1475 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DAPK2| Gene Symbol: DAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521414.1 | XP_011519716.1 | 11 | 2417 | ||
| XM_017022044.1 | XP_016877533.1 | 11 | 2413 | |||
| GenBank mRNA | AK094290.1 | - | 1 | 1475 | ||