siRNA Details
| Gene Symbol: DAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521414.1 | XP_011519716.1 | 11 | 1381 | ||
| XM_017022044.1 | XP_016877533.1 | 11 | 1377 | |||
| GenBank mRNA | AK094290.1 | - | 1 | 439 | ||
| BC114951.1 | - | 10 | 1996 | 40005391 | ||

DAPK2
Genomic Map