siRNA Details
| Gene Symbol: TMEM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110503.2 | NP_001103973.1 | 7 | 1889 | ||
| GenBank mRNA | AK023569.1 | - | 1 | 235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM87A| Gene Symbol: TMEM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110503.2 | NP_001103973.1 | 7 | 1889 | ||
| GenBank mRNA | AK023569.1 | - | 1 | 235 | ||