siRNA Details
| Gene Symbol: LOC105377832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037882.2 | - | 1 | 1656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105377832| Gene Symbol: LOC105377832 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037882.2 | - | 1 | 1656 | ||