siRNA Details
| Gene Symbol: LOC102724612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125825.1 | 2 | 1608 | |||
| GenBank mRNA | BC040302.1 | - | 2 | 1609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724612| Gene Symbol: LOC102724612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125825.1 | 2 | 1608 | |||
| GenBank mRNA | BC040302.1 | - | 2 | 1609 | ||