siRNA Details
Gene Symbol: LOC102724612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125825.1 | 2 | 1559 | |||
GenBank mRNA | BC040302.1 | - | 2 | 1560 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102724612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125825.1 | 2 | 1559 | |||
GenBank mRNA | BC040302.1 | - | 2 | 1560 |