siRNA Details
| Gene Symbol: SLC39A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525901.1 | XP_011524203.1 | 2 | 1337 | ||
| GenBank mRNA | AK097938.1 | - | 1 | 1721 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC39A6| Gene Symbol: SLC39A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525901.1 | XP_011524203.1 | 2 | 1337 | ||
| GenBank mRNA | AK097938.1 | - | 1 | 1721 | ||