siRNA Details
| Gene Symbol: TMSB4XP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC090885.1 | - | 1 | 517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMSB4XP1| Gene Symbol: TMSB4XP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC090885.1 | - | 1 | 517 | ||