siRNA Details
| Gene Symbol: NAAA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005262925.3 | XP_005262982.1 | 9 | 2537 | ||
| XM_017008029.1 | XP_016863518.1 | 10 | 2566 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK025371.1 | - | 1 | 139 | Chr. 4 - 75914379 - 75916406 [-] on Build GRCh38 | |

NAAA
Genomic Map