siRNA Details
| Gene Symbol: MGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534559.1 | XP_011532861.1 | 1 | 142 | ||
| XM_011534562.1 | XP_011532864.1 | 1 | 70 | |||
| XM_011534563.2 | XP_011532865.1 | 1 | 125 | |||
| XM_017009486.1 | XP_016864975.1 | 1 | 125 | |||
| GenBank mRNA | AK057194.1 | - | 1 | 31 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK124428.1 | - | 1 | 31 | Chr. 5 - 180813053 - 180815541 [-] on Build GRCh38 | |

MGAT1
Genomic Map