siRNA Details
| Gene Symbol: GATA2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024653.1 | - | 1 | 656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GATA2-AS1| Gene Symbol: GATA2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024653.1 | - | 1 | 656 | ||