siRNA Details
| Gene Symbol: ROPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK026656.1 | - | 1 | 439 | ||
| Gene Symbol: LOC105369214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011514190.2 | XP_011512492.1 | 1 | 745 | ||

ROPN1L
Genomic Map