siRNA Details
| Gene Symbol: SLC9A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284351.2 | NP_001271280.1 | 17 | 3889 | ||
| NM_004174.3 | NP_004165.2 | 17 | 3916 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK023178.1 | - | 1 | 984 | Chr. 5 - 470511 - 473063 [-] on Build GRCh38 | |

SLC9A3
Genomic Map