siRNA Details
| Gene Symbol: DIO3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037351.1 | - | 1 | 2240 | ||
| BC065701.1 | - | 3 | 1014 | |||
| BC111049.1 | - | 2 | 831 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DIO3OS| Gene Symbol: DIO3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037351.1 | - | 1 | 2240 | ||
| BC065701.1 | - | 3 | 1014 | |||
| BC111049.1 | - | 2 | 831 | |||