siRNA Details
| Gene Symbol: DIO3OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC037351.1 | - | 1 | 2476 | ||
| BC065701.1 | - | 3 | 1250 | |||
| BC111049.1 | - | 2 | 1067 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ636731.1 | - | 1 | 162 | Chr. 14 - 101557478 - 101558100 [-] on Build GRCh38 | |

DIO3OS
Genomic Map