siRNA Details
| Gene Symbol: LINC01279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109779.1 | 1 | 3940 | |||
| GenBank mRNA | AK126393.1 | - | 1 | 2346 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG545874.1 | - | 1 | 69 | Chr. 3 - 112597326 - 112598098 [-] on Build GRCh38 | |

LINC01279
Genomic Map