siRNA Details
| Gene Symbol: LINC01279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109779.1 | 1 | 1713 | |||
| GenBank mRNA | AK126393.1 | - | 1 | 119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01279| Gene Symbol: LINC01279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109779.1 | 1 | 1713 | |||
| GenBank mRNA | AK126393.1 | - | 1 | 119 | ||