siRNA Details
| Gene Symbol: ZBTB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 5929 | ||
| NM_001164343.2 | NP_001157815.1 | 12 | 6391 | |||
| NM_001164344.2 | NP_001157816.1 | 8 | 6355 | |||
| NM_001164345.2 | NP_001157817.1 | 7 | 6309 | |||
| NM_001164346.2 | NP_001157818.1 | 6 | 5948 | |||
| NM_001164347.2 | NP_001157819.1 | 6 | 5970 | |||
| NM_015642.5 | NP_056457.3 | 10 | 6153 | |||
| NR_121662.1 | 7 | 4411 | ||||
| GenBank mRNA | AK022228.1 | - | 1 | 1770 | ||

ZBTB20
Genomic Map