siRNA Details
| Gene Symbol: ZBTB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 5443 | ||
| NM_001164343.2 | NP_001157815.1 | 12 | 5905 | |||
| NM_001164344.2 | NP_001157816.1 | 8 | 5869 | |||
| NM_001164345.2 | NP_001157817.1 | 7 | 5823 | |||
| NM_001164346.2 | NP_001157818.1 | 6 | 5462 | |||
| NM_001164347.2 | NP_001157819.1 | 6 | 5484 | |||
| NM_015642.5 | NP_056457.3 | 10 | 5667 | |||
| NR_121662.1 | 7 | 3925 | ||||
| GenBank mRNA | AK022228.1 | - | 1 | 1284 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DN991835.1 | - | 1 | 97 | Chr. 3 - 114335476 - 114336057 [-] on Build GRCh38 | |

ZBTB20
Genomic Map