siRNA Details
| Gene Symbol: LOC101928068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111907.1 | 2 | 1828 | |||
| GenBank mRNA | AK126649.1 | - | 1 | 845 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928068| Gene Symbol: LOC101928068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111907.1 | 2 | 1828 | |||
| GenBank mRNA | AK126649.1 | - | 1 | 845 | ||