siRNA Details
| Gene Symbol: PKI55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037701.1 | 1 | 1459 | |||
| GenBank mRNA | AK125369.1 | - | 1 | 1303 | ||
| AL137534.1 | - | 1 | 763 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKI55| Gene Symbol: PKI55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037701.1 | 1 | 1459 | |||
| GenBank mRNA | AK125369.1 | - | 1 | 1303 | ||
| AL137534.1 | - | 1 | 763 | |||