siRNA Details
| Gene Symbol: HHLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097806.1 | - | 1 | 1493 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB213093.1 | - | 1 | 508 | Chr. 3 - 108388581 - 108389134 [+] on Build GRCh38 | |

HHLA2
Genomic Map