siRNA Details
| Gene Symbol: ART3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008206.1 | XP_016863695.1 | 10 | 3210 | ||
| XM_017008207.1 | XP_016863696.1 | 9 | 3180 | |||
| XM_017008208.1 | XP_016863697.1 | 12 | 3723 | |||
| GenBank mRNA | AK129914.1 | - | 1 | 896 | ||

ART3
Genomic Map