siRNA Details
| Gene Symbol: ART3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017008206.1 | XP_016863695.1 | 10 | 2893 | ||
| XM_017008207.1 | XP_016863696.1 | 9 | 2863 | |||
| XM_017008208.1 | XP_016863697.1 | 12 | 3406 | |||
| GenBank mRNA | AK129914.1 | - | 1 | 579 | ||

ART3
Genomic Map