siRNA Details
| Gene Symbol: LINC00884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK091527.1 | - | 4 | 2524 | ||
| AK094122.1 | - | 1 | 1762 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ215608.1 | - | 1 | 278 | Chr. 3 - 194517828 - 194518334 [+] on Build GRCh38 | |

LINC00884
Genomic Map