siRNA Details
| Gene Symbol: HOXB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005257286.2 | XP_005257343.1 | 1 | 3694 | ||
| XM_017024564.1 | XP_016880053.1 | 1 | 3694 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK025239.1 | - | 2 | 130 | Chr. 17 - 48617095 - 48622528 [-] on Build GRCh38 | |

HOXB8
Genomic Map