siRNA Details
| Gene Symbol: ZKSCAN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242894.1 | NP_001229823.1 | 7 | 4326 | ||
| NM_001242895.1 | NP_001229824.1 | 5 | 3745 | |||
| NM_024493.3 | NP_077819.2 | 6 | 4209 | |||
| XM_005249423.3 | XP_005249480.1 | 5 | 4375 | |||
| XM_006715215.2 | XP_006715278.1 | 6 | 4107 | |||
| XM_006715216.2 | XP_006715279.1 | 6 | 4122 | |||
| XM_006715218.3 | XP_006715281.1 | 5 | 3673 | |||
| XM_017011320.1 | XP_016866809.1 | 7 | 4203 | |||

ZKSCAN3
Genomic Map