siRNA Details
Gene Symbol: TOR1AIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199260.1 | NP_001186189.1 | 7 | 6530 | ||
NM_145034.4 | NP_659471.1 | 6 | 6111 | |||
XM_005244939.4 | XP_005244996.1 | 7 | 6503 | |||
XM_017000474.1 | XP_016855963.1 | 8 | 6648 | |||
XM_017000475.1 | XP_016855964.1 | 6 | 6372 | |||
XM_017000476.1 | XP_016855965.1 | 3 | 5812 | |||
GenBank mRNA | AK002036.1 | - | 1 | 1433 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX099455.1 | - | 1 | 62 | Chr. 1 - 179841137 - 179841830 [-] on Build GRCh38 |
