siRNA Details
| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199260.1 | NP_001186189.1 | 7 | 7300 | ||
| NM_145034.4 | NP_659471.1 | 6 | 6881 | |||
| XM_005244939.4 | XP_005244996.1 | 7 | 7273 | |||
| XM_017000474.1 | XP_016855963.1 | 8 | 7418 | |||
| XM_017000475.1 | XP_016855964.1 | 6 | 7142 | |||
| XM_017000476.1 | XP_016855965.1 | 3 | 6582 | |||
| GenBank mRNA | AK002036.1 | - | 1 | 2203 | ||

TOR1AIP2
Genomic Map