siRNA Details
| Gene Symbol: ATXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC011026.1 | - | 2 | 1212 | ||
| BC014548.1 | - | 3 | 1281 | |||
| BC039236.1 | - | 3 | 1205 | |||
| BC047894.1 | - | 3 | 1310 | |||
| BC063120.1 | - | 3 | 1305 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATXN1| Gene Symbol: ATXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC011026.1 | - | 2 | 1212 | ||
| BC014548.1 | - | 3 | 1281 | |||
| BC039236.1 | - | 3 | 1205 | |||
| BC047894.1 | - | 3 | 1310 | |||
| BC063120.1 | - | 3 | 1305 | |||