siRNA Details
| Gene Symbol: LGALS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB209391.1 | - | 1 | 1669 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK091776.1 | - | 1 | 1827 | Chr. 14 - 55132966 - 55135154 [+] on Build GRCh38 | |

LGALS3
Genomic Map