siRNA Details
| Gene Symbol: LOC100127947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY240960.1 | - | 2 | 1826 | ||
| Gene Symbol: SH3D21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017002340.1 | XP_016857829.1 | 18 | 3823 | ||
| XM_017002341.1 | XP_016857830.1 | 17 | 3817 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN268700.1 | - | 1 | 615 | Chr. 1 - 36328595 - 36329252 [+] on Build GRCh38 | |

LOC100127947
Genomic Map