siRNA Details
| Gene Symbol: TTN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038271.1 | 5 | 1209 | |||
| GenBank mRNA | AK093733.1 | - | 5 | 1176 | ||
| AK094031.1 | - | 1 | 740 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTN-AS1| Gene Symbol: TTN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038271.1 | 5 | 1209 | |||
| GenBank mRNA | AK093733.1 | - | 5 | 1176 | ||
| AK094031.1 | - | 1 | 740 | |||