siRNA Details
| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006719437.2 | XP_006719500.1 | 17 | 3756 | ||
| XM_017019385.1 | XP_016874874.1 | 18 | 3837 | |||
| GenBank mRNA | AK093735.1 | - | 1 | 807 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEP83| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006719437.2 | XP_006719500.1 | 17 | 3756 | ||
| XM_017019385.1 | XP_016874874.1 | 18 | 3837 | |||
| GenBank mRNA | AK093735.1 | - | 1 | 807 | ||