siRNA Details
| Gene Symbol: CEP83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006719437.2 | XP_006719500.1 | 17 | 4841 | ||
| XM_017019385.1 | XP_016874874.1 | 18 | 4922 | |||
| GenBank mRNA | AK093735.1 | - | 1 | 1892 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA702661.1 | - | 1 | 76 | Chr. 12 - 94304026 - 94304495 [-] on Build GRCh38 | |

CEP83
Genomic Map